Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AURKA SIRNA Assays ›
          Gene Symbol:
          AURKA AURKAPS1
          Gene Name:
          aurora kinase A
          aurora kinase A pseudogene 1
          Gene Aliases:
          AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
          AurAps1, STK6P
          Chromosome Location:
          Chr.20: 56369389 - 56392337 on Build GRCh38
          Chromosome Location:
          Chr.1: 220266179 - 220267715 on Build GRCh38
          UniGene ID:
          Hs.250822
          Hs.654849
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 427

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AURKA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001323303.1 NP_001310232.1 4 758
          NM_001323304.1 NP_001310233.1 4 521
          NM_001323305.1 NP_001310234.1 5 631
          NM_003600.3 NP_003591.2 3 648
          NM_198433.2 NP_940835.1 5 856
          NM_198434.2 NP_940836.1 4 547
          NM_198435.2 NP_940837.1 3 437
          NM_198436.2 NP_940838.1 4 533
          NM_198437.2 NP_940839.1 3 423
          XM_017028034.1 XP_016883523.1 5 978
          XM_017028035.1 XP_016883524.1 3 338
          GenBank mRNA AF008551.1 - 2 248
          AF011468.1 - 3 504
          AK312776.1 - 3 315
          AL711075.1 - 3 516
          BC001280.1 - 4 380 3456017
          BC002499.2 - 3 376 3051177
          BC006423.1 - 3 273 3532438
          BC027464.1 - 4 415 4449474
          BF029182.1 - 304
          BM458030.1 - 4 448
          BP224535.1 - 3 437
          BQ435059.1 - 3 269
          BU171303.1 - 3 369
          BU542363.1 - 5 708
          D84212.1 - 4 417
          Gene Symbol: AURKAPS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_001587.1 1 47

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          meiotic spindle organization
          protein phosphorylation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          spindle organization
          mitotic spindle organization
          spindle assembly involved in female meiosis I
          mitotic nuclear division
          mitotic centrosome separation
          anterior/posterior axis specification
          anaphase-promoting complex-dependent catabolic process
          regulation of protein stability
          negative regulation of protein binding
          response to estradiol
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of cytokinesis
          histone-serine phosphorylation
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          positive regulation of mitotic nuclear division
          regulation of centrosome cycle
          protein autophosphorylation
          cell division
          centrosome localization
          protein localization to centrosome
          positive regulation of oocyte maturation
          regulation of signal transduction by p53 class mediator
          neuron projection extension
          protein kinase activity
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          ubiquitin protein ligase binding
          histone serine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0