Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZP3 SIRNA Assays ›
Gene Symbol:
Zp3
Gene Name:
zona pellucida glycoprotein 3 (sperm receptor)
Gene Aliases:
Zp-3
Chromosome Location:
Chr.12: 23735225 - 23742190 on Build Rnor_6.0
UniGene ID:
Rn.10892
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
55538
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Zp3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053762.1
NP_446214.1
1
166
GenBank mRNA
BC127488.1
-
1
166
7308805
D78482.1
-
1
166
Y10823.1
-
1
159
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of type IV hypersensitivity
blastocyst formation
humoral immune response mediated by circulating immunoglobulin
positive regulation of leukocyte migration
positive regulation of humoral immune response
manganese ion transport
intracellular protein transport
binding of sperm to zona pellucida
positive regulation of phosphatidylinositol biosynthetic process
positive regulation of interferon-gamma production
positive regulation of interleukin-4 production
intracellular signal transduction
egg coat formation
positive regulation of T cell proliferation
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
phosphatidylinositol-mediated signaling
oocyte development
positive regulation of inflammatory response
positive regulation of protein kinase B signaling
protein kinase C signaling
calcium ion transmembrane transport
manganese ion transmembrane transport
positive regulation of calcium ion import
positive regulation of acrosome reaction
negative regulation of binding of sperm to zona pellucida
positive regulation of acrosomal vesicle exocytosis
positive regulation of ovarian follicle development
positive regulation of antral ovarian follicle growth
signal transducer activity
manganese ion transmembrane transporter activity
store-operated calcium channel activity
carbohydrate binding
acrosin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0