Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172041.2 | NP_742038.2 | 2 | 338 | ||
NM_172042.1 | NP_742039.1 | 2 | 326 | |||
XM_006250433.2 | XP_006250495.1 | 2 | 549 | |||
XM_006250434.2 | XP_006250496.1 | 2 | 928 | |||
GenBank mRNA | AF325671.1 | - | 2 | 326 | ||
AF385402.1 | - | 2 | 433 | |||
AY555072.1 | - | 2 | 289 | |||
AY555073.1 | - | 2 | 310 | |||
AY695826.1 | - | 2 | 283 | |||
AY727922.1 | - | 2 | 281 | |||
DQ403851.1 | - | 2 | 338 |