Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR3C1 SIRNA Assays ›
          Gene Symbol:
          Nr3c1
          Gene Name:
          nuclear receptor subfamily 3, group C, member 1
          Gene Aliases:
          GR, Grl-1, Grl1
          Chromosome Location:
          Chr.18: 39410545 - 39519416 on Build GRCm38
          UniGene ID:
          Mm.129481
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61881

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nr3c1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008173.3 NP_032199.3 1 144
          GenBank mRNA AK037733.1 - 1 24
          AK155200.1 - 2 253
          AK156323.1 - 2 247
          BC008154.1 - 2 455
          BC016462.1 - 2 254
          BC129912.1 - 1 149 40111802
          BC129913.1 - 1 149 40111803
          BY232938.1 - 2 345
          DQ504162.1 - 1 131
          HM236293.1 - 1 131
          X04435.1 - 1 149
          X13358.1 - 1 131
          X13359.1 - 1 131

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001018074, NM_001018075, NM_001018076, NM_001204261, NM_000176, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of gluconeogenesis
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          glucocorticoid metabolic process
          regulation of glucose metabolic process
          positive regulation of glutamate secretion
          chromatin modification
          adrenal gland development
          negative regulation of synaptic plasticity
          regulation of glucocorticoid biosynthetic process
          regulation of cell proliferation
          maternal behavior
          glucocorticoid receptor signaling pathway
          negative regulation of vascular permeability
          steroid hormone mediated signaling pathway
          glucocorticoid mediated signaling pathway
          positive regulation of neuron apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          response to arsenic-containing substance
          chromatin-mediated maintenance of transcription
          mammary gland duct morphogenesis
          positive regulation of dendritic spine development
          positive regulation of cell growth involved in cardiac muscle cell development
          cellular response to steroid hormone stimulus
          cellular response to dexamethasone stimulus
          negative regulation of glucocorticoid mediated signaling pathway
          positive regulation of glucocorticoid receptor signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II regulatory region DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          glucocorticoid receptor activity
          steroid binding
          protein binding
          transcription factor binding
          zinc ion binding
          lipid binding
          Hsp70 protein binding
          receptor tyrosine kinase binding
          heat shock protein binding
          protein complex binding
          glucocorticoid-activated RNA polymerase II transcription factor binding transcription factor activity
          hormone binding
          protein homodimerization activity
          sequence-specific DNA binding
          metal ion binding
          protein heterodimerization activity
          protein dimerization activity
          Hsp90 protein binding
          steroid hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-q8kbf:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0