Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010499.4 | NP_034629.3 | 1 | 1043 | ||
GenBank mRNA | AK032692.1 | - | 1041 | |||
AK144601.1 | - | 1 | 1040 | |||
AK150860.1 | - | 1 | 1038 | |||
AK152747.1 | - | 1 | 1038 | |||
AK170433.1 | - | 1040 | ||||
AK172332.1 | - | 1 | 985 | |||
BC002067.1 | - | 1 | 1028 | 3590296 | ||
M31042.1 | - | 1 | 1042 | |||
M59821.1 | - | 1 | 1045 |