Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRPM4 SIRNA Assays ›
          Gene Symbol:
          TRPM4
          Gene Name:
          transient receptor potential cation channel subfamily M member 4
          Gene Aliases:
          LTrpC4, PFHB1B, TRPM4B, hTRPM4
          Chromosome Location:
          Chr.19: 49157741 - 49211841 on Build GRCh38
          UniGene ID:
          Hs.467101
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7595

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRPM4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195227.1 NP_001182156.1 8, 9 1166
          NM_001321281.1 NP_001308210.1 6, 7 839
          NM_001321282.1 NP_001308211.1 7, 8 1003
          NM_001321283.1 NP_001308212.1 6, 7 828
          NM_017636.3 NP_060106.2 8, 9 1166
          XM_005259017.1 XP_005259074.1 3, 4 372
          GenBank mRNA AF497623.1 - 8, 9 1130
          AJ575813.1 - 8, 9 1099
          AK000048.1 - 6, 7 859
          AK292862.1 - 8, 9 1119
          AK307897.1 - 6, 7 841
          AY046396.1 - 6, 7 758
          AY297044.1 - 6, 7 754
          AY297045.1 - 8, 9 1110
          AY297046.1 - 8, 9 1110
          BC132727.1 - 8, 9 1093 40146749
          BC144123.1 - 8, 9 1093

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          dendritic cell chemotaxis
          regulation of T cell cytokine production
          positive regulation of cytosolic calcium ion concentration
          positive regulation of cell proliferation
          positive regulation of heart rate
          protein sumoylation
          negative regulation of bone mineralization
          sodium ion transmembrane transport
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          vasoconstriction
          positive regulation of fat cell differentiation
          negative regulation of osteoblast differentiation
          positive regulation of vasoconstriction
          cardiac conduction
          calcium ion transmembrane transport
          positive regulation of canonical Wnt signaling pathway
          positive regulation of atrial cardiac muscle cell action potential
          positive regulation of adipose tissue development
          positive regulation of regulation of vascular smooth muscle cell membrane depolarization
          calcium activated cation channel activity
          calcium channel activity
          sodium channel activity
          calmodulin binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0