Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Oser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025699.2 | NP_079975.2 | 3 | 217 | ||
GenBank mRNA | AK006969.1 | - | 3 | 260 | ||
AK009708.1 | - | 3 | 233 | |||
AK014323.1 | - | 3 | 236 | |||
AK088622.1 | - | 3 | 236 | |||
AK139692.1 | - | 389 | ||||
AK153499.1 | - | 3 | 254 | |||
BC005579.1 | - | 3 | 217 | 3968876 |