Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM10921 SIRNA Assays ›
          Gene Symbol:
          Gm10921 Gm10922 Gm14345 Gm14346 Gm14347 Gm14351 Gm14374 Gm21950 Gm21951 Gm2799 Gm2825 Gm2863 Gm2913 Gm2927 Gm2933 Gm3701 Gm3706 Gm3750 Gm3763 Gm5925 Gmcl1l
          Gene Name:
          predicted gene 10921
          predicted gene 10922
          predicted gene 14345
          predicted gene 14346
          predicted gene 14347
          predicted gene 14351
          predicted gene 14374
          predicted gene, 21950
          predicted gene, 21951
          predicted gene 2799
          predicted gene 2825
          predicted gene 2863
          predicted gene 2913
          predicted gene 2927
          predicted gene 2933
          predicted gene 3701
          predicted gene 3706
          predicted gene 3750
          predicted gene 3763
          germ cell-less homolog 1 family pseudogene
          germ cell-less homolog 1 (Drosophila)-like
          Gene Aliases:
          ENSMUSG00000078215, OTTMUSG00000016420
          ENSMUSG00000078216, OTTMUSG00000016437
          OTTMUSG00000016405
          OTTMUSG00000016406
          OTTMUSG00000016407
          OTTMUSG00000016411
          OTTMUSG00000016453
          Mgclh
          Mgclh
          -
          GMCL1P1, Gclh, Gmcl1l
          -
          -
          -
          -
          -
          -
          -
          -
          EG546185
          1700017G21Rik, Gclh, Gmcl2
          Chromosome Location:
          Chr.X: 4289286 - 4291245 on Build GRCm38
          Chromosome Location:
          Chr.X: 4370636 - 4372595 on Build GRCm38
          Chromosome Location:
          Chr.X: 3700233 - 3702192 on Build GRCm38
          Chromosome Location:
          Chr.X: 3441731 - 3443690 on Build GRCm38
          Chromosome Location:
          Chr.X: 4196576 - 4198535 on Build GRCm38
          Chromosome Location:
          Chr.X: 3750931 - 3752885 on Build GRCm38
          Chromosome Location:
          Chr.X: 5669067 - 5671026 on Build GRCm38
          Chromosome Location:
          Chr.X: 3076920 - 3078823 on Build GRCm38
          Chromosome Location:
          Chr.X: 32726137 - 32728057 on Build GRCm38
          Chromosome Location:
          Chr.X: 32278394 - 32280342 on Build GRCm38
          Chromosome Location:
          Chr.X: 32973897 - 32975846 on Build GRCm38
          Chromosome Location:
          Chr.X: 33313338 - 33315288 on Build GRCm38
          Chromosome Location:
          Chr.X: 33575387 - 33577346 on Build GRCm38
          Chromosome Location:
          Chr.X: 33657140 - 33659082 on Build GRCm38
          Chromosome Location:
          Chr.X: 33700649 - 33702349 on Build GRCm38
          Chromosome Location:
          Chr.X: 3955043 - 3957002 on Build GRCm38
          Chromosome Location:
          Chr.X: 4037674 - 4039622 on Build GRCm38
          Chromosome Location:
          Chr.X: 4800411 - 4802359 on Build GRCm38
          Chromosome Location:
          Chr.X: 4952135 - 4954077 on Build GRCm38
          Chromosome Location:
          Chr.X: 4515589 - 4517543 on Build GRCm38
          Chromosome Location:
          Chr.X: 32560055 - 32562009 on Build GRCm38
          UniGene ID:
          Mm.457966
          Mm.457962
          Mm.457961
          Mm.443264
          Mm.457960
          -
          Mm.457958
          Mm.443264
          Mm.337329
          Mm.479861
          Mm.337329
          Mm.460713
          Mm.489578
          -
          Mm.465135
          Mm.489579
          Mm.489575
          -
          Mm.427352
          Mm.359482
          Mm.379203
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 82343

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm10921
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085553.1 NP_001079022.1 1 219
          Gene Symbol: Gm10922
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085554.1 NP_001079023.1 1 219
          Gene Symbol: Gm14345
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085545.1 NP_001079014.1 1 219
          Gene Symbol: Gm14346
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085551.1 NP_001079020.1 1 219
          Gene Symbol: Gm14347
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085543.1 NP_001079012.1 1 219
          Gene Symbol: Gm14351
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085552.2 NP_001079021.1 1 219
          Gene Symbol: Gm14374
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085523.1 NP_001078992.1 1 219
          Gene Symbol: Gm21950
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270667.1 NP_001257596.1 1 174
          Gene Symbol: Gm21951
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270669.1 NP_001257598.1 1 191
          GenBank mRNA AB055854.1 - 1 191
          Gene Symbol: Gm2799
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001168334.1 NP_001161806.1 1 219
          Gene Symbol: Gm2825
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001168337.1 NP_001161809.1 1 219
          Gene Symbol: Gm2863
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099333.3 NP_001092803.3 1 219
          Gene Symbol: Gm2913
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243015.1 NP_001229944.1 1 219
          Gene Symbol: Gm2927
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282034.1 NP_001268963.1 1 219
          Gene Symbol: Gm2933
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145038.1 NP_001138510.1 1 187
          GenBank mRNA BC115962.1 - 1 187 40057634
          Gene Symbol: Gm3701
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243011.1 NP_001229940.1 1 219
          Gene Symbol: Gm3706
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243001.1 NP_001229930.1 1 219
          Gene Symbol: Gm3750
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099643.2 NP_001093113.1 1 219
          Gene Symbol: Gm3763
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243024.1 NP_001229953.1 1 219
          GenBank mRNA BC089532.1 - 1 219 6704091
          Gene Symbol: Gm5925
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC115856.1 - 1 187 40057635
          Gene Symbol: Gmcl1l
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_027955.3 NP_082231.2 1 219
          GenBank mRNA AB055856.1 - 177
          AK006065.1 - 1 200

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-fxmhk:80/100.66.79.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0