Search
Search

FAM96B| Gene Symbol: FAM96B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016062.3 | NP_057146.1 | 2 | 239 | ||
| NR_024525.2 | 1 | 395 | ||||
| NR_046109.1 | 2 | 305 | ||||
| GenBank mRNA | AF151886.1 | - | 2 | 215 | ||
| AF161467.1 | - | 2 | 204 | |||
| BC001733.1 | - | 2 | 199 | 3353705 | ||
| BC005023.1 | - | 2 | 202 | 3632169 | ||
| BI598300.1 | - | 1 | 390 | |||
| BI603322.1 | - | 2 | 269 | |||
| BM091095.1 | - | 1 | 319 | |||
| BQ955804.1 | - | 2 | 171 | |||
| CD674578.1 | - | 2 | 223 | |||
| DA270099.1 | - | 1 | 396 | |||
| DR155511.1 | - | 1 | 436 | |||