Search
Search

PCDHAC1| Gene Symbol: PCDHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018898.3 | NP_061721.2 | 1 | 1059 | ||
| NM_031882.3 | NP_114088.2 | 1 | 1059 | |||
| GenBank mRNA | AF152303.1 | - | 1 | 883 | ||
| AF152473.1 | - | 1 | 883 | |||
| AJ007608.1 | - | 1 | 883 | |||
| BC136774.1 | - | 1 | 904 | 9020764 | ||
| BC136785.1 | - | 1 | 904 | 9020775 | ||