Search
Search

CHKA| Gene Symbol: CHKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277.2 | NP_001268.2 | 11 | 1486 | ||
| NM_212469.1 | NP_997634.1 | 10 | 1432 | |||
| XM_017017147.1 | XP_016872636.1 | 10 | 1370 | |||
| XM_017017148.1 | XP_016872637.1 | 10 | 2842 | |||
| GenBank mRNA | BE260889.1 | - | 1 | 26 | ||
| BI195927.1 | - | 1 | 47 | |||
| D10704.1 | - | 11 | 1296 | |||