Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADCYAP1 SIRNA Assays ›
          Gene Symbol:
          ADCYAP1
          Gene Name:
          adenylate cyclase activating polypeptide 1
          Gene Aliases:
          PACAP
          Chromosome Location:
          Chr.18: 904342 - 912172 on Build GRCh38
          UniGene ID:
          Hs.531719
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS141151
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADCYAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099733.1 NP_001093203.1 2 153
          NM_001117.4 NP_001108.2 1 225
          XM_005258081.3 XP_005258138.1 3 552
          GenBank mRNA AK294233.1 - 3 481
          AK313050.1 - 2 153
          BC093837.1 - 1 105 7939682
          BC101803.1 - 1 105 8069309
          DA145895.1 - 1 312
          DC320719.1 - 2 153
          DN991692.1 - 1 132
          S83513.1 - 1 35

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          behavioral fear response
          histamine secretion
          negative regulation of acute inflammatory response to antigenic stimulus
          negative regulation of acute inflammatory response to non-antigenic stimulus
          activation of adenylate cyclase activity
          positive regulation of cytosolic calcium ion concentration
          neuropeptide signaling pathway
          cell-cell signaling
          female pregnancy
          regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of cell proliferation
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          negative regulation of muscle cell apoptotic process
          positive regulation of neuron projection development
          sensory perception of pain
          cAMP-mediated signaling
          pituitary gland development
          neuron projection development
          positive regulation of interleukin-6 production
          regulation of protein localization
          negative regulation of GTPase activity
          response to starvation
          negative regulation of potassium ion transport
          positive regulation of GTPase activity
          response to ethanol
          negative regulation of cell cycle
          positive regulation of protein kinase activity
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          ATP metabolic process
          positive regulation of synaptic transmission, glutamatergic
          regulation of postsynaptic membrane potential
          positive regulation of growth hormone secretion
          negative regulation of glial cell proliferation
          positive regulation of ERK1 and ERK2 cascade
          regulation of oligodendrocyte progenitor proliferation
          cellular response to glucocorticoid stimulus
          positive regulation of chemokine (C-C motif) ligand 5 production
          positive regulation of somatostatin secretion
          receptor signaling protein activity
          receptor binding
          neuropeptide hormone activity
          protein binding
          pituitary adenylate cyclase activating polypeptide activity
          pituitary adenylate cyclase-activating polypeptide receptor binding
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0