Search
Search

TMEM107| Gene Symbol: TMEM107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032354.3 | NP_115730.2 | 4 | 419 | ||
| NM_183065.2 | NP_898888.1 | 4 | 401 | |||
| GenBank mRNA | AF311338.1 | - | 4 | 475 | ||
| AK027813.1 | - | 3 | 1518 | |||
| AK127891.1 | - | 2 | 1609 | |||
| AY358525.1 | - | 4 | 379 | |||
| BC070231.1 | - | 4 | 411 | 6617879 | ||
| BC127649.1 | - | 4 | 378 | 40118154 | ||
| BM920977.1 | - | 4 | 372 | |||
| BU852346.1 | - | 4 | 414 | |||