Search
Search

GPR45| Gene Symbol: GPR45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007227.3 | NP_009158.3 | 1 | 1198 | ||
| GenBank mRNA | BC066879.1 | - | 1 | 1198 | 6971748 | |
| BC066880.1 | - | 1 | 1198 | 6971750 | ||
| BC067455.1 | - | 1 | 1198 | 6971749 | ||
| EU883573.1 | - | 1 | 1082 | |||
Set Membership: |
|