Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTX2 SIRNA Assays ›
Gene Symbol:
DTX2
DTX2P1-UPK3BP1-PMS2P11
Gene Name:
deltex E3 ubiquitin ligase 2
DTX2P1-UPK3BP1-PMS2P11 readthrough, transcribed pseudogene
Gene Aliases:
RNF58
PMS2L11
Chromosome Location:
Chr.7: 76461655 - 76505995 on Build GRCh38
Chromosome Location:
Chr.7: 76980822 - 77023761 on Build GRCh38
UniGene ID:
Hs.187058
Hs.675888
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS174312
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102594.1
NP_001096064.1
4
1093
NM_001102595.1
NP_001096065.1
3
928
NM_001102596.1
NP_001096066.1
2
657
NM_020892.2
NP_065943.2
5
1213
XM_005250131.1
XP_005250188.1
5
1104
XM_005250132.1
XP_005250189.1
3
847
XM_011515772.1
XP_011514074.1
5
1275
XM_011515773.2
XP_011514075.1
5
1702
XM_011515774.1
XP_011514076.1
5
1028
XM_011515775.1
XP_011514077.1
4
1122
XM_017011723.1
XP_016867212.1
4
1151
XM_017011724.1
XP_016867213.1
4
1582
XM_017011725.1
XP_016867214.1
4
1000
XM_017011726.1
XP_016867215.1
4
1093
XM_017011727.1
XP_016867216.1
3
907
XM_017011728.1
XP_016867217.1
2
746
XM_017011729.1
XP_016867218.1
2
746
XM_017011730.1
XP_016867219.1
2
746
GenBank mRNA
AB040961.1
-
4
1015
AK023924.1
-
3
869
AK293226.1
-
3
682
AY225124.1
-
2
657
AY225125.1
-
2
657
BC008856.2
-
3
848
4107018
BC018555.1
-
5
1167
4341568
BC026059.2
-
3
848
4099111
BC093079.1
-
4
981
30512572
DQ010329.1
-
5
1167
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_023383.1
1
188
GenBank mRNA
BP230837.1
-
1
188
U38980.1
-
1
80
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
Notch signaling pathway
protein ubiquitination
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0