Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013447.3 | NP_038475.2 | 14 | 1884 | ||
XM_011527948.2 | XP_011526250.1 | 13 | 1513 | |||
XM_011527949.2 | XP_011526251.1 | 13 | 1480 | |||
XM_011527951.2 | XP_011526253.1 | 12 | 1366 | |||
XM_011527953.2 | XP_011526255.1 | 11 | 1320 | |||
XM_011527954.2 | XP_011526256.1 | 10 | 2051 | |||
XM_011527955.2 | XP_011526257.1 | 8 | 930 | |||
XM_017026726.1 | XP_016882215.1 | 12 | 1482 | |||
GenBank mRNA | AF114491.1 | - | 13 | 1497 | ||
AK222891.1 | - | 13 | 1482 | |||
AK291776.1 | - | 13 | 1630 | |||
AK298690.1 | - | 8 | 735 | |||
BC082991.1 | - | 13 | 1441 | |||
BC127004.1 | - | 14 | 1880 | 40108332 | ||
BC127005.1 | - | 14 | 1880 | 40108333 | ||
CR936819.1 | - | 8 | 861 |
Set Membership: |
G-protein coupled receptor |