Search
Search

RTCB| Gene Symbol: RTCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014306.4 | NP_055121.1 | 10 | 1365 | ||
| GenBank mRNA | AF155658.1 | - | 10 | 1313 | ||
| AF161466.1 | - | 10 | 1324 | |||
| AK297826.1 | - | 10 | 1242 | |||
| AK312503.1 | - | 10 | 1325 | |||
| AL050255.1 | - | 10 | 1309 | |||
| AL137272.1 | - | 3 | 311 | |||
| BC000151.1 | - | 10 | 1303 | 2984824 | ||
| BC002970.1 | - | 10 | 1303 | 3533883 | ||
| BC010308.1 | - | 10 | 1315 | 3451576 | ||
| BC016707.1 | - | 10 | 1330 | 4102148 | ||
| CR456450.1 | - | 10 | 1280 | |||
| CR457175.1 | - | 10 | 1234 | |||