Search
Search

HECW2| Gene Symbol: HECW2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304840.1 | NP_001291769.1 | 15, 16 | 3188 | ||
| NM_020760.2 | NP_065811.1 | 17, 18 | 3522 | |||
| XM_006712646.3 | XP_006712709.1 | 17, 18 | 3580 | |||
| XM_006712648.3 | XP_006712711.1 | 16, 17 | 3513 | |||
| XM_017004563.1 | XP_016860052.1 | 17, 18 | 3559 | |||
| GenBank mRNA | AB037722.1 | - | 17, 18 | 3522 | ||
| BC117194.1 | - | 17, 18 | 3438 | 40125745 | ||
| BC117198.1 | - | 17, 18 | 3438 | 40125749 | ||
| CR749424.1 | - | 15, 16 | 3286 | |||