Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MROH1 SIRNA Assays ›
Gene Symbol:
MROH1
Gene Name:
maestro heat like repeat family member 1
Gene Aliases:
HEATR7A
Chromosome Location:
Chr.8: 144148016 - 144261940 on Build GRCh38
UniGene ID:
Hs.443139
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS186543
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MROH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099280.2
NP_001092750.1
8
887
NM_001099281.2
NP_001092751.1
7
767
NM_001288814.1
NP_001275743.1
7
774
NM_032450.2
NP_115826.2
8
887
XM_011517270.2
XP_011515572.1
8
1025
XM_011517271.2
XP_011515573.1
8
1024
XM_011517272.2
XP_011515574.1
8
1025
XM_011517273.2
XP_011515575.1
8
1023
XM_011517274.2
XP_011515576.1
8
1023
XM_011517275.2
XP_011515577.1
8
1023
XM_011517276.2
XP_011515578.1
8
1051
XM_011517277.1
XP_011515579.1
8
887
XM_011517278.1
XP_011515580.1
7
767
XM_011517279.2
XP_011515581.1
7
931
XM_011517281.1
XP_011515583.1
7
940
XM_011517282.1
XP_011515584.1
6
865
XM_011517283.2
XP_011515585.1
8
846
XM_011517285.1
XP_011515587.1
7
767
XM_011517286.2
XP_011515588.1
8
1019
XM_017013802.1
XP_016869291.1
8
1023
XM_017013803.1
XP_016869292.1
8
1022
XM_017013804.1
XP_016869293.1
8
887
XM_017013805.1
XP_016869294.1
7
767
XM_017013806.1
XP_016869295.1
5
502
XM_017013807.1
XP_016869296.1
8
1020
GenBank mRNA
AB058736.1
-
7
779
AK093563.1
-
8
886
AK295740.1
-
7
767
AL834309.1
-
7
771
BC040742.1
-
5
514
BC057824.1
-
8
867
BC067855.1
-
8
867
BC092486.1
-
7
747
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline