Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DMKN SIRNA Assays ›
          Gene Symbol:
          DMKN
          Gene Name:
          dermokine
          Gene Aliases:
          UNQ729, ZD52F10
          Chromosome Location:
          Chr.19: 35497217 - 35513678 on Build GRCh38
          UniGene ID:
          Hs.417795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS190172
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DMKN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126056.2 NP_001119528.2 3, 4 862
          NM_001126057.2 NP_001119529.2 3, 4 862
          NM_001126058.2 NP_001119530.2 3, 4 862
          NM_001190347.1 NP_001177276.1 3, 4 862
          NM_001190348.1 NP_001177277.1 3, 4 862
          NM_001190349.1 NP_001177278.1 3, 4 862
          NM_033317.4 NP_201574.3 3, 4 862
          XM_011527494.2 XP_011525796.1 3, 4 882
          XM_011527495.2 XP_011525797.1 3, 4 882
          XM_011527496.2 XP_011525798.1 3, 4 882
          XM_011527497.2 XP_011525799.1 3, 4 882
          XM_011527498.2 XP_011525800.1 3, 4 882
          XM_011527499.2 XP_011525801.1 3, 4 882
          XM_011527500.2 XP_011525802.1 3, 4 882
          XM_011527501.2 XP_011525803.1 3, 4 882
          XM_011527502.2 XP_011525804.1 3, 4 882
          XM_011527503.2 XP_011525805.1 3, 4 882
          XM_011527504.2 XP_011525806.1 3, 4 882
          XM_011527505.2 XP_011525807.1 3, 4 882
          XM_011527506.2 XP_011525808.1 3, 4 882
          XM_011527507.2 XP_011525809.1 3, 4 882
          XM_011527508.2 XP_011525810.1 3, 4 882
          XM_011527509.2 XP_011525811.1 3, 4 882
          XM_011527510.2 XP_011525812.1 3, 4 882
          XM_011527511.2 XP_011525813.1 3, 4 882
          XM_011527512.2 XP_011525814.1 3, 4 882
          GenBank mRNA AY358412.1 - 3, 4 862
          AY622965.1 - 3, 4 679
          AY789695.1 - 3, 4 685
          AY789696.1 - 3, 4 685
          AY789697.1 - 3, 4 685
          BC035311.1 - 3, 4 856 4752921
          EF158319.1 - 3, 4 685
          EF158320.1 - 3, 4 686
          EF158321.1 - 3, 4 679
          EF158322.1 - 3, 4 684

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cornified envelope assembly
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0