Search
Search

Zfp638| Gene Symbol: Zfp638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166371.1 | NP_001159843.1 | 2 | 1304 | ||
| NM_008717.3 | NP_032743.2 | 2 | 1314 | |||
| GenBank mRNA | AK043029.1 | - | 2 | 1286 | ||
| AK157791.1 | - | 2 | 1295 | |||
| AK163476.1 | - | 1 | 910 | |||
| AK163955.1 | - | 2 | 1266 | |||
| AK165189.1 | - | 2 | 1285 | |||
| BC019223.1 | - | 2 | 1286 | |||
| BC076615.1 | - | 2 | 1267 | 30640388 | ||
| D83033.1 | - | 2 | 1300 | |||