Search
Search

Scg2| Gene Symbol: Scg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009129.2 | NP_033155.1 | 2 | 952 | ||
| GenBank mRNA | AK002953.1 | - | 1 | 449 | ||
| BC014717.1 | - | 2 | 921 | |||
| BC048249.1 | - | 2 | 852 | 6493827 | ||
| BE863515.1 | - | 1 | 22 | |||