Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011766.5 | NP_035896.1 | 4 | 900 | ||
GenBank mRNA | AF107306.1 | - | 4 | 421 | ||
AF118845.1 | - | 4 | 670 | |||
AF125166.1 | - | 4 | 901 | |||
AK046521.1 | - | 4 | 908 | |||
BC059241.1 | - | 4 | 800 | 6408975 | ||
EF514219.1 | - | 4 | 1723 |