Search
Search

Megf9| Gene Symbol: Megf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172694.2 | NP_766282.1 | 5 | 1368 | ||
| GenBank mRNA | AK035987.1 | - | 5 | 1369 | ||
| AK039319.1 | - | 5 | 1369 | |||
| AK122378.1 | - | 1359 | ||||
| BC088994.1 | - | 5 | 1277 | 6734479 | ||
| BC138128.1 | - | 5 | 1307 | 8861147 | ||
| BC138129.1 | - | 5 | 1307 | 8861148 | ||