Search
Search

Rdh10| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133832.3 | NP_598593.1 | 6 | 1095 | ||
| GenBank mRNA | AF456767.1 | - | 6 | 1203 | ||
| AK014244.1 | - | 2 | 763 | |||
| AK149656.1 | - | 6 | 1166 | |||
| BC019796.1 | - | 6 | 1095 | 4923005 | ||
| BC038334.1 | - | 1 | 203 | |||
| BC052546.1 | - | 1 | 207 | |||