Search
Search

Zfp46| Gene Symbol: Zfp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009557.3 | NP_033583.2 | 3 | 750 | ||
| GenBank mRNA | AK051031.1 | - | 3 | 723 | ||
| AK053652.1 | - | 3 | 753 | |||
| AK136405.1 | - | 3 | 659 | |||
| AK148255.1 | - | 3 | 699 | |||
| AK173240.1 | - | 3 | 737 | |||
| BC006587.1 | - | 3 | 417 | 3587306 | ||
| BC092398.1 | - | 1 | 60 | |||
| M98502.1 | - | 1 | 273 | |||