Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081264.1 | NP_001074733.1 | 14 | 1378 | ||
GenBank mRNA | AK157903.1 | - | 14 | 1340 | ||
AK171887.1 | - | 14 | 1331 | |||
BC044875.1 | - | 4 | 213 | |||
BC050854.1 | - | 14 | 1343 | 6332345 | ||
BC080867.1 | - | 13 | 3335 |