Search
Search

Pcbp4| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021567.5 | NP_067542.3 | 11 | 973 | ||
| GenBank mRNA | AF176328.1 | - | 11 | 859 | ||
| AK004580.1 | - | 11 | 922 | |||
| AK005383.1 | - | 11 | 922 | |||
| AK077632.1 | - | 11 | 923 | |||
| AK082005.1 | - | 11 | 923 | |||
| AK179994.1 | - | 1 | 17 | |||
| AK181953.1 | - | 17 | ||||
| AK188369.1 | - | 1 | 17 | |||
| AK188891.1 | - | 1 | 17 | |||
| AK216715.1 | - | 1 | 17 | |||
| BC010694.1 | - | 11 | 862 | 2654291 | ||