Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148413.3 | NP_680779.3 | 533 | |||
GenBank mRNA | AK018511.1 | - | 532 | |||
AY101368.1 | - | 376 |