Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAX SIRNA Assays ›
Gene Symbol:
MAX
LOC100506321
Gene Name:
MYC associated factor X
uncharacterized LOC100506321
Gene Aliases:
bHLHd4
-
Chromosome Location:
Chr.14: 65006101 - 65102695 on Build GRCh38
Chromosome Location:
Chr.14: 65089918 - 65094311 on Build GRCh38
UniGene ID:
Hs.285354
Hs.711267
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
VHS40426
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271068.1
NP_001257997.1
2
260
NM_001271069.1
NP_001257998.1
2
260
NM_001320415.1
NP_001307344.1
3
287
NM_002382.4
NP_002373.3
3
287
NM_145112.2
NP_660087.1
2
260
NM_145113.2
NP_660088.1
3
287
NM_145114.2
NP_660089.1
3
287
NM_197957.3
NP_932061.1
3
287
NR_073137.1
2
206
NR_073138.1
2
206
XM_011536773.2
XP_011535075.1
3
275
XM_017021312.1
XP_016876801.1
2
246
XM_017021313.1
XP_016876802.1
3
166
GenBank mRNA
AI652319.1
-
2
197
AI655674.1
-
2
197
AI769390.1
-
2
451
AI917842.1
-
2
197
AK290130.1
-
2
191
AK290929.1
-
3
260
AK292189.1
-
3
197
AK292630.1
-
3
135
AK303412.1
-
2
160
AL833643.1
-
3
263
AY353088.1
-
2
55
BC003525.1
-
2
170
3607261
BC004516.2
-
3
213
3937573
BC013669.1
-
3
252
4155088
BC025685.1
-
2
207
5205305
BC027924.1
-
2
217
5309524
BC036092.1
-
3
233
5299106
BF038005.1
-
3
298
CR542191.1
-
2
55
DB451269.1
-
2
208
DB482510.1
-
2
233
HY018878.1
-
2
133
M64240.1
-
3
109
X60287.1
-
3
103
Gene Symbol: LOC100506321
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_045122.1
3
377
GenBank mRNA
BI825681.1
-
3
328
DB082809.1
-
3
377
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to starvation
negative regulation of gene expression
response to insulin
response to axon injury
neuron apoptotic process
retina development in camera-type eye
cellular response to peptide hormone stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription coactivator activity
protein binding
protein complex binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0