Search
Search

| Gene Symbol: DPY19L2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003551.1 | 3 | 654 | |||
| GenBank mRNA | BC031277.1 | - | 3 | 611 | ||
| BC047471.1 | - | 3 | 602 | |||
| BX537815.1 | - | 3 | 654 | |||
| DQ287934.1 | - | 3 | 654 | |||
| Gene Symbol: DPY19L2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC023575.2 | - | 4 | 2337 | ||
| BC068601.1 | - | 3 | 2046 | |||