Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026945.2 | 3 | 654 | |||
NR_108024.1 | 3 | 635 | ||||
NR_108025.1 | 2 | 564 | ||||
GenBank mRNA | AK093725.1 | - | 3 | 1350 | ||
BC093717.1 | - | 1 | 127 | |||
BC111994.1 | - | 1 | 127 | |||
BG494977.1 | - | 2 | 536 | |||
BQ053612.1 | - | 3 | 616 | |||
CD518314.1 | - | 3 | 599 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI857138.1 | - | 1 | 28 | Chr. 5 - 170333485 - 170334407 [+] on Build GRCh38 |