Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10223
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
13, 14
1129
NM_001001568.1
NP_001001568.1
11, 12
980
NM_001001569.1
NP_001001569.1
10, 11
925
NM_001001570.1
NP_001001570.1
13, 14
1150
NM_001001571.1
NP_001001571.1
12, 13
1058
NM_001001572.1
NP_001001572.1
10, 11
936
NM_001001573.1
NP_001001573.1
11, 12
1116
NM_001001574.1
NP_001001574.1
12, 13
1051
NM_001001575.1
NP_001001575.1
9, 10
881
NM_001001576.1
NP_001001576.1
9, 10
840
NM_001001577.1
NP_001001577.1
10, 11
959
NM_001001578.1
NP_001001578.1
11, 12
1003
NM_001001579.1
NP_001001579.1
11, 12
996
NM_001001580.1
NP_001001580.1
8, 9
796
NM_001001581.1
NP_001001581.1
12, 13
1160
NM_001001582.1
NP_001001582.1
13, 14
1238
NM_001001583.1
NP_001001583.1
13, 14
1231
NM_001001584.2
NP_001001584.1
12, 13
1059
NM_001001585.1
NP_001001585.1
12, 13
1183
NM_001315533.1
NP_001302462.1
14, 15
1205
NM_002606.2
NP_002597.1
14, 15
1309
XM_011529598.2
XP_011527900.1
14, 15
1231
XM_011529600.2
XP_011527902.1
14, 15
1282
XM_017028366.1
XP_016883855.1
12, 13
1047
GenBank mRNA
AF048837.1
-
14, 15
1306
AF067223.1
-
14, 15
1258
AF067224.1
-
13, 14
1077
AF067225.1
-
11, 12
919
AF067226.1
-
10, 11
864
AK001165.1
-
11, 12
911
AK001307.1
-
13, 14
1100
AK127770.1
-
4, 5
2660
AK294112.1
-
11, 12
946
AK308753.1
-
8, 9
785
AK314679.1
-
14, 15
1282
AY196299.1
-
13, 14
1123
AY196300.1
-
12, 13
1031
AY196301.1
-
10, 11
909
AY196302.1
-
11, 12
1089
AY196303.1
-
12, 13
1024
AY196304.1
-
9, 10
854
AY196305.1
-
9, 10
813
AY196306.1
-
10, 11
932
AY196307.1
-
11, 12
976
AY196308.1
-
11, 12
969
AY196309.1
-
8, 9
769
AY196310.1
-
12, 13
1133
AY196311.1
-
13, 14
1211
AY196312.1
-
13, 14
1204
AY196313.1
-
12, 13
1031
AY196314.1
-
12, 13
1156
AY242121.1
-
12, 13
995
AY701187.1
-
14, 15
1202
BC009047.1
-
13, 14
1103
3874635
BT007016.1
-
13, 14
1066
KU178246.1
-
13, 14
1066
KU178247.1
-
14, 15
1246
KU178248.1
-
10, 11
862
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline