Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290720.1 | NP_001277649.1 | 6 | 826 | ||
NM_001290721.1 | NP_001277650.1 | 5 | 633 | |||
NM_207228.3 | NP_997111.1 | 6 | 693 | |||
GenBank mRNA | AF530050.3 | - | 5 | 638 | ||
AK030254.1 | - | 6 | 826 | |||
AK149243.1 | - | 2 | 2590 | |||
AK163138.1 | - | 6 | 640 | |||
AK170053.1 | - | 6 | 716 | |||
AY618264.1 | - | 5 | 638 | |||
BC053407.1 | - | 5 | 616 | |||
BC066782.1 | - | 6 | 690 | 6705127 |