Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172723.4 | NP_766311.2 | 6 | 625 | ||
GenBank mRNA | AK076754.1 | - | 5 | 611 | ||
AK076768.1 | - | 6 | 382 | |||
BC056934.1 | - | 5 | 351 | |||
BC079634.1 | - | 5 | 419 | |||
BC144874.1 | - | 5 | 370 |