Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172742.2 | NP_766330.2 | 11 | 1225 | ||
GenBank mRNA | BC041672.1 | - | 11 | 1197 | ||
BC055074.1 | - | 9 | 889 | 30052208 | ||
BC058196.1 | - | 4 | 327 | |||
CF734341.1 | - | 6 | 604 |