Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARP1 SIRNA Assays ›
          Gene Symbol:
          PARP1
          Gene Name:
          poly(ADP-ribose) polymerase 1
          Gene Aliases:
          ADPRT, ADPRT 1, ADPRT1, ARTD1, PARP, PARP-1, PPOL, pADPRT-1
          Chromosome Location:
          Chr.1: 226360691 - 226408100 on Build GRCh38
          UniGene ID:
          Hs.177766
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1098
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PARP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001618.3 NP_001609.2 17 2545
          GenBank mRNA AK125650.1 - 3 902
          AK225654.1 - 17 2510
          AK303340.1 - 17 2472
          AK312339.1 - 17 2461
          BC037545.1 - 17 2518 5193735
          J03473.1 - 17 2469
          M17081.1 - 8 1054
          M18112.1 - 17 2513
          M32721.1 - 17 2533

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00242302_m1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          nucleotide-excision repair, DNA damage recognition
          nucleotide-excision repair, DNA duplex unwinding
          double-strand break repair via homologous recombination
          lagging strand elongation
          DNA repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          double-strand break repair
          transcription from RNA polymerase II promoter
          protein ADP-ribosylation
          cellular response to DNA damage stimulus
          mitochondrion organization
          transforming growth factor beta receptor signaling pathway
          positive regulation of cardiac muscle hypertrophy
          protein autoprocessing
          protein sumoylation
          signal transduction involved in regulation of gene expression
          macrophage differentiation
          mitochondrial DNA metabolic process
          cellular response to insulin stimulus
          nucleotide-excision repair, DNA incision
          cellular response to oxidative stress
          protein modification process
          DNA damage response, detection of DNA damage
          mitochondrial DNA repair
          positive regulation of transcription from RNA polymerase II promoter
          DNA ligation involved in DNA repair
          positive regulation of SMAD protein import into nucleus
          protein poly-ADP-ribosylation
          global genome nucleotide-excision repair
          regulation of cellular protein localization
          negative regulation of telomere maintenance via telomere lengthening
          ATP generation from poly-ADP-D-ribose
          positive regulation of transcription regulatory region DNA binding
          DNA binding
          DNA ligase (ATP) activity
          NAD+ ADP-ribosyltransferase activity
          protein binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          identical protein binding
          histone deacetylase binding
          poly(A) RNA binding
          protein N-terminus binding
          NAD binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0