Search
Search

Map7d1| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144941.3 | NP_659190.3 | 6 | 1053 | ||
| GenBank mRNA | AK122462.1 | - | 1 | 939 | ||
| AK208379.1 | - | 1 | 61 | |||
| BC023677.1 | - | 6 | 1028 | 5354497 | ||
| BM944941.1 | - | 4 | 344 | |||
| CV563343.1 | - | 5 | 624 | |||