Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPK10 SIRNA Assays ›
Gene Symbol:
MAPK10
Gene Name:
mitogen-activated protein kinase 10
Gene Aliases:
JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
Chromosome Location:
Chr.4: 86012296 - 86594117 on Build GRCh38
UniGene ID:
Hs.125503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11161
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318067.1
NP_001304996.1
9
944
NM_001318068.1
NP_001304997.1
9
1039
NM_001318069.1
NP_001304998.1
10
1509
NM_002753.4
NP_002744.1
10
1045
NM_138980.3
NP_620446.1
10
1536
NM_138982.3
NP_620448.1
10
1509
XM_005263129.2
XP_005263186.1
10
1606
XM_005263130.2
XP_005263187.1
10
1002
XM_005263131.2
XP_005263188.1
10
1403
XM_005263135.3
XP_005263192.1
10
1403
XM_006714268.2
XP_006714331.1
10
1276
XM_006714269.2
XP_006714332.1
10
1403
XM_011532117.2
XP_011530419.1
10
1109
XM_011532118.2
XP_011530420.1
10
1202
XM_011532120.2
XP_011530422.1
10
1172
XM_011532121.2
XP_011530423.1
10
1038
XM_017008419.1
XP_016863908.1
11
1299
XM_017008420.1
XP_016863909.1
10
1733
XM_017008421.1
XP_016863910.1
10
1329
XM_017008422.1
XP_016863911.1
7
757
XM_017008423.1
XP_016863912.1
10
1021
XM_017008424.1
XP_016863913.1
10
1356
XM_017008425.1
XP_016863914.1
10
1029
XM_017008426.1
XP_016863915.1
10
1357
XM_017008427.1
XP_016863916.1
10
1430
XM_017008428.1
XP_016863917.1
10
1172
XM_017008429.1
XP_016863918.1
10
1733
XM_017008430.1
XP_016863919.1
10
1111
XM_017008431.1
XP_016863920.1
10
1329
XM_017008432.1
XP_016863921.1
10
1002
XM_017008433.1
XP_016863922.1
9
1358
XM_017008434.1
XP_016863923.1
10
1386
XM_017008435.1
XP_016863924.1
10
1276
XM_017008436.1
XP_016863925.1
10
1760
XM_017008437.1
XP_016863926.1
10
1038
XM_017008438.1
XP_016863927.1
10
1029
XM_017008439.1
XP_016863928.1
10
1357
XM_017008440.1
XP_016863929.1
10
1430
XM_017008441.1
XP_016863930.1
9
1358
XM_017008442.1
XP_016863931.1
12
1751
XM_017008443.1
XP_016863932.1
12
1402
XM_017008444.1
XP_016863933.1
12
1724
XM_017008445.1
XP_016863934.1
9
1233
XM_017008446.1
XP_016863935.1
7
1925
XM_017008447.1
XP_016863936.1
9
1371
XM_017008448.1
XP_016863937.1
9
1344
XM_017008449.1
XP_016863938.1
9
1371
XM_017008450.1
XP_016863939.1
9
1344
XM_017008451.1
XP_016863940.1
9
1170
XM_017008452.1
XP_016863941.1
9
1371
GenBank mRNA
AK057723.1
-
5
831
AK091104.1
-
2
775
AK124791.1
-
10
1761
AK225974.1
-
10
1090
AK294125.1
-
6
560
BC022492.1
-
10
1342
BC035057.1
-
10
1354
5261221
BC051731.1
-
10
1155
4838495
BC065516.1
-
5
506
5721954
U07620.1
-
10
1046
U34819.1
-
9
890
U34820.1
-
9
914
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
protein phosphorylation
signal transduction
JNK cascade
JUN phosphorylation
response to light stimulus
Fc-epsilon receptor signaling pathway
regulation of circadian rhythm
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
JUN kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0