Search
Search

MAP2K6| Gene Symbol: MAP2K6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002758.3 | NP_002749.2 | 9, 10 | 1027 | ||
| XM_005257515.2 | XP_005257572.1 | 9, 10 | 1091 | |||
| XM_005257516.2 | XP_005257573.1 | 9, 10 | 964 | |||
| XM_006721975.3 | XP_006722038.1 | 9, 10 | 992 | |||
| XM_011525025.2 | XP_011523327.1 | 9, 10 | 1378 | |||
| XM_011525026.2 | XP_011523328.1 | 9, 10 | 839 | |||
| XM_011525027.2 | XP_011523329.1 | 9, 10 | 754 | |||
| GenBank mRNA | AB451252.1 | - | 9, 10 | 739 | ||
| AB451376.1 | - | 9, 10 | 739 | |||
| AK290747.1 | - | 9, 10 | 1039 | |||
| BC012009.1 | - | 9, 10 | 937 | 4499772 | ||
| D87905.1 | - | 9, 10 | 739 | |||
| U39064.1 | - | 9, 10 | 975 | |||
| U39065.1 | - | 9, 10 | 1024 | |||
| U39656.1 | - | 9, 10 | 1079 | |||
| U39657.1 | - | 9, 10 | 1079 | |||
| U49732.1 | - | 9, 10 | 984 | |||
| X96757.1 | - | 9, 10 | 1011 | |||