Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSMC6 SIRNA Assays ›
          Gene Symbol:
          PSMC6
          Gene Name:
          proteasome 26S subunit, ATPase 6
          Gene Aliases:
          CADP44, HEL-S-73, P44, SUG2, p42
          Chromosome Location:
          Chr.14: 52707163 - 52727998 on Build GRCh38
          UniGene ID:
          Hs.156171
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s11385
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PSMC6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002806.3 NP_002797.3 12 972
          XM_017021472.1 XP_016876961.1 12 2475
          GenBank mRNA AF006305.1 - 12 945
          AK313670.1 - 12 940
          BC005390.1 - 12 950 3996906
          BT006843.1 - 12 930
          CR456709.1 - 12 930
          D78275.1 - 12 946
          FJ224299.1 - 12 946

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs01652481_g1

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          ubiquitin-dependent protein catabolic process
          regulation of cellular amino acid metabolic process
          ER-associated ubiquitin-dependent protein catabolic process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          positive regulation of proteasomal protein catabolic process
          protein binding
          ATP binding
          ATPase activity
          TBP-class protein binding
          protein binding, bridging
          proteasome-activating ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-hrqtj:80/100.66.76.20:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline