Search
Search

Cyb5r4| Gene Symbol: Cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024195.2 | NP_077157.2 | 12 | 1261 | ||
| GenBank mRNA | AF338818.1 | - | 11 | 1009 | ||
| AK012855.1 | - | 11 | 900 | |||
| AK034878.1 | - | 12 | 1205 | |||
| AK078682.1 | - | 5 | 467 | |||
| AK090159.1 | - | 12 | 1263 | |||
| AK135261.1 | - | 11 | 912 | |||
| AK157312.1 | - | 11 | 1265 | |||
| AK167436.1 | - | 11 | 959 | |||
| AK169937.1 | - | 11 | 1259 | |||
| AK169995.1 | - | 11 | 1256 | |||
| AK172252.1 | - | 11 | 1214 | |||
| AK216438.1 | - | 2 | 161 | |||
| AK217521.1 | - | 2 | 161 | |||
| AY321368.1 | - | 10 | 1033 | |||
| BC002170.1 | - | 5 | 461 | |||
| BC025438.1 | - | 12 | 1272 | 5038563 | ||