Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172947.3 | NP_766535.1 | 4 | 502 | ||
GenBank mRNA | AK011126.1 | - | 4 | 489 | ||
AK075668.1 | - | 4 | 504 | |||
AK081685.1 | - | 477 | ||||
AK132889.1 | - | 4 | 499 | |||
AK151118.1 | - | 505 | ||||
BC027784.1 | - | 4 | 515 | 5343888 |