Search
Search

Wnt11| Gene Symbol: Wnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080401.1 | NP_536326.1 | 4 | 1076 | ||
| XM_006229718.2 | XP_006229780.1 | 4 | 928 | |||
| XM_006229719.2 | XP_006229781.1 | 5 | 1377 | |||
| XM_006229720.2 | XP_006229782.1 | 5 | 2299 | |||
| XM_008759667.1 | XP_008757889.1 | 4 | 649 | |||
| GenBank mRNA | AB073722.1 | - | 2 | 296 | ||