Search
Search

Klc1| Gene Symbol: Klc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081972.1 | NP_001075441.1 | 13 | 1616 | ||
| NM_001081973.1 | NP_001075442.1 | 13 | 1639 | |||
| NM_001081974.1 | NP_001075443.1 | 13 | 1625 | |||
| XM_006240621.2 | XP_006240683.1 | 13 | 2209 | |||
| XM_006240623.2 | XP_006240685.1 | 13 | 2210 | |||
| XM_006240625.2 | XP_006240687.1 | 13 | 2105 | |||
| XM_006240626.2 | XP_006240688.1 | 13 | 2104 | |||
| XM_006240627.2 | XP_006240689.1 | 13 | 2105 | |||
| XM_006240629.2 | XP_006240691.1 | 13 | 2104 | |||
| GenBank mRNA | M75146.1 | - | 13 | 1616 | ||
| M75147.1 | - | 13 | 1639 | |||
| M75148.1 | - | 13 | 1625 | |||