Search
Search

| Gene Symbol: Olr1610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000094.1 | NP_001000094.1 | 1 | 392 | ||
| XM_008770652.1 | XP_008768874.1 | 1 | 392 | |||
| Gene Symbol: Olr1611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751458.3 | XP_003751506.1 | 1 | 430 | ||