Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SORBS2 SIRNA Assays ›
          Gene Symbol:
          SORBS2
          Gene Name:
          sorbin and SH3 domain containing 2
          Gene Aliases:
          ARGBP2, PRO0618
          Chromosome Location:
          Chr.4: 185585444 - 185956716 on Build GRCh38
          UniGene ID:
          Hs.481342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16089

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SORBS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145670.1 NP_001139142.1 10 1453
          NM_001145671.2 NP_001139143.1 10 1026
          NM_001145672.1 NP_001139144.1 9 823
          NM_001145673.1 NP_001139145.1 10 1550
          NM_001145674.1 NP_001139146.1 3 472
          NM_001145675.1 NP_001139147.1 4 517
          NM_001270771.1 NP_001257700.1 12 1507
          NM_003603.6 NP_003594.3 10 1340
          NM_021069.4 NP_066547.1 9 1287
          XM_005263307.1 XP_005263364.1 12 2144
          XM_005263308.1 XP_005263365.1 11 1502
          XM_005263310.1 XP_005263367.1 11 1433
          XM_005263311.1 XP_005263368.1 11 1502
          XM_005263312.1 XP_005263369.1 10 1340
          XM_005263313.1 XP_005263370.1 10 1457
          XM_006714388.1 XP_006714451.1 11 1433
          XM_006714390.1 XP_006714453.1 10 1409
          XM_017008738.1 XP_016864227.1 13 2618
          XM_017008739.1 XP_016864228.1 14 2921
          XM_017008740.1 XP_016864229.1 13 2614
          XM_017008741.1 XP_016864230.1 13 2618
          XM_017008742.1 XP_016864231.1 12 2568
          XM_017008743.1 XP_016864232.1 13 2549
          XM_017008744.1 XP_016864233.1 13 2828
          XM_017008745.1 XP_016864234.1 12 2768
          XM_017008746.1 XP_016864235.1 13 2618
          XM_017008747.1 XP_016864236.1 13 2759
          XM_017008748.1 XP_016864237.1 13 2618
          XM_017008749.1 XP_016864238.1 13 2180
          XM_017008750.1 XP_016864239.1 13 2618
          XM_017008751.1 XP_016864240.1 13 2618
          XM_017008752.1 XP_016864241.1 13 2618
          XM_017008753.1 XP_016864242.1 13 2618
          XM_017008754.1 XP_016864243.1 12 2573
          XM_017008755.1 XP_016864244.1 10 1583
          XM_017008756.1 XP_016864245.1 9 1286
          XM_017008757.1 XP_016864246.1 3 472
          XM_017008758.1 XP_016864247.1 10 1452
          XM_017008759.1 XP_016864248.1 10 2179
          XM_017008760.1 XP_016864249.1 10 1409
          XM_017008761.1 XP_016864250.1 13 2618
          XM_017008762.1 XP_016864251.1 11 1502
          XM_017008763.1 XP_016864252.1 10 1452
          XM_017008764.1 XP_016864253.1 11 1433
          XM_017008765.1 XP_016864254.1 12 1736
          XM_017008766.1 XP_016864255.1 10 1409
          XM_017008767.1 XP_016864256.1 11 1712
          XM_017008768.1 XP_016864257.1 10 1630
          XM_017008769.1 XP_016864258.1 10 1388
          XM_017008770.1 XP_016864259.1 9 1364
          XM_017008771.1 XP_016864260.1 9 1290
          XM_017008772.1 XP_016864261.1 10 1340
          XM_017008773.1 XP_016864262.1 11 1643
          XM_017008774.1 XP_016864263.1 4 917
          XM_017008775.1 XP_016864264.1 12 2573
          GenBank mRNA AB018320.1 - 9 939
          AF049884.1 - 10 1304
          AF049885.1 - 9 669
          AK056628.1 - 12 1125
          AK056758.1 - 3 472
          AK225327.1 - 10 1419
          AK225812.1 - 10 1550
          AK293400.1 - 9 823
          AK295742.1 - 10 858
          AK296461.1 - 3 249
          AK304653.1 - 4 253
          BC011883.1 - 10 993 3677070
          DA836871.1 - 3 472
          DC421607.1 - 4 516

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          cell adhesion
          biological_process
          cell migration
          cell growth involved in cardiac muscle cell development
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal adaptor activity
          structural constituent of muscle
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0