Search
Search

TOMM70| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014820.4 | NP_055635.3 | 9, 10 | 1909 | ||
| GenBank mRNA | AB018262.1 | - | 9, 10 | 1551 | ||
| AK075080.1 | - | 4, 5 | 402 | |||
| AK075102.1 | - | 4, 5 | 519 | |||
| AK302796.1 | - | 10, 11 | 1453 | |||
| BC003633.2 | - | 9, 10 | 1575 | 3605218 | ||
| BC052994.1 | - | 9, 10 | 1892 | 5549394 | ||
| BC065555.1 | - | 9, 10 | 1906 | |||