Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36 SIRNA Assays ›
          Gene Symbol:
          ANKRD36
          Gene Name:
          ankyrin repeat domain 36
          Gene Aliases:
          UNQ2430
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          UniGene ID:
          Hs.541894
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s200088

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 41, 42 2825
          XM_006712514.3 XP_006712577.1 41, 42 3135
          XM_011511130.1 XP_011509432.1 41, 42 3135
          XM_011511131.1 XP_011509433.1 41, 42 3135
          XM_017004009.1 XP_016859498.1 41, 42 3135
          XM_017004010.1 XP_016859499.1 41, 42 3135
          XM_017004011.1 XP_016859500.1 41, 42 3135
          XM_017004012.1 XP_016859501.1 41, 42 3135
          XM_017004013.1 XP_016859502.1 39, 40 3033
          XM_017004014.1 XP_016859503.1 41, 42 3135
          XM_017004015.1 XP_016859504.1 39, 40 3033
          XM_017004016.1 XP_016859505.1 41, 42 3135
          XM_017004017.1 XP_016859506.1 39, 40 3033
          XM_017004018.1 XP_016859507.1 39, 40 3033
          XM_017004019.1 XP_016859508.1 39, 40 3033
          XM_017004020.1 XP_016859509.1 39, 40 3033
          XM_017004021.1 XP_016859510.1 41, 42 3135
          XM_017004022.1 XP_016859511.1 39, 40 3033
          XM_017004023.1 XP_016859512.1 41, 42 3135
          XM_017004024.1 XP_016859513.1 40, 41 2997
          XM_017004025.1 XP_016859514.1 41, 42 3135
          XM_017004026.1 XP_016859515.1 41, 42 3135
          XM_017004027.1 XP_016859516.1 41, 42 3135
          XM_017004028.1 XP_016859517.1 37, 38 2931
          XM_017004029.1 XP_016859518.1 37, 38 2931
          XM_017004031.1 XP_016859520.1 41, 42 3135
          XM_017004032.1 XP_016859521.1 37, 38 2931
          XM_017004033.1 XP_016859522.1 41, 42 3135
          XM_017004034.1 XP_016859523.1 41, 42 3135
          XM_017004035.1 XP_016859524.1 37, 38 2931
          XM_017004036.1 XP_016859525.1 41, 42 3135
          XM_017004037.1 XP_016859526.1 41, 42 3135
          XM_017004038.1 XP_016859527.1 41, 42 3135
          XM_017004039.1 XP_016859528.1 35, 36 2829
          XM_017004041.1 XP_016859530.1 41, 42 3135
          XM_017004042.1 XP_016859531.1 41, 42 3135
          XM_017004043.1 XP_016859532.1 41, 42 3135
          XM_017004045.1 XP_016859534.1 41, 42 3135
          XM_017004046.1 XP_016859535.1 39, 40 2890
          XM_017004047.1 XP_016859536.1 38, 39 2822
          XM_017004048.1 XP_016859537.1 37, 38 2749
          XM_017004049.1 XP_016859538.1 39, 40 2924
          XM_017004050.1 XP_016859539.1 24, 25 1328
          XM_017004051.1 XP_016859540.1 22, 23 1542
          GenBank mRNA BC128262.1 - 7, 8 912
          DA952980.1 - 4, 5 199

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zxk44:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0