Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003563.1 | 1 | 107 | |||
GenBank mRNA | AK096724.1 | - | 3 | 333 | ||
AK292695.1 | - | 2 | 198 | |||
BC072398.1 | - | 3 | 383 | |||
BC131538.1 | - | 1 | 107 |
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 33 | 3935 | ||
XM_005261634.1 | XP_005261691.1 | 32 | 3842 | |||
XM_005261635.1 | XP_005261692.1 | 33 | 3935 | |||
XM_011530226.1 | XP_011528528.1 | 33 | 3935 | |||
XM_017028829.1 | XP_016884318.1 | 32 | 3869 | |||
XM_017028830.1 | XP_016884319.1 | 19 | 2091 | |||
GenBank mRNA | AB210002.1 | - | 33 | 3905 | ||
AF012872.1 | - | 32 | 3674 | |||
AK302424.1 | - | 33 | 3863 | |||
AK314781.1 | - | 5 | 589 | |||
BC018120.1 | - | 3 | 252 | |||
BC053654.1 | - | 2 | 228 | |||
L36151.1 | - | 1 | 221 |
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003700.1 | 3 | 374 | |||
GenBank mRNA | AK126860.1 | - | 1 | 2338 | ||
AK298744.1 | - | 3 | 395 | |||
AK299734.1 | - | 3 | 239 | |||
BC020225.1 | - | 3 | 372 | |||
BC139841.1 | - | 1 | 95 | |||
BX452846.2 | - | 3 | 386 | |||
CR456378.1 | - | 1 | 113 | |||
DA463001.1 | - | 3 | 318 |